KEY001136423 Key Organics / BIONET

2-(4-fluorophenoxy)-1-methyl-1H-indole-3-carbaldehyde

  • MDL number MFCD01315828
  • Cas Number : 338416-70-9
  • Molecular Weight : 269.275

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139 - 141
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72130

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3N-335S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart