KEY001136436 Key Organics / BIONET

1-methyl-2-(2,4,5-trichlorophenoxy)-1H-indole-3-carbaldehyde

  • MDL number MFCD01315836
  • Cas Number : 338416-88-9
  • Molecular Weight : 354.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160 - 163
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72144

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3N-352S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart