KEY001136864 Key Organics / BIONET

6-{[(4-chlorophenyl)formohydrazido]carbonyl}cyclohex-3-ene-1-carboxylic acid

  • MDL number MFCD02186201
  • Cas Number : 451485-61-3
  • Molecular Weight : 322.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180 - 182
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72677

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-0254_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart