KEY001136927 Key Organics / BIONET

4-(1,1-dioxo-1lambda6-thiomorpholin-4-yl)benzoic acid

  • MDL number MFCD02186426
  • Cas Number : 451485-62-4
  • Molecular Weight : 255.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 308 - 311
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72759

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-0668_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart