KEY001137060 Key Organics / BIONET

4-methyl-2-(methylsulfanyl)pyrimidine-5-carboxylic acid

  • MDL number MFCD02187391
  • Cas Number : 98276-75-6
  • Molecular Weight : 184.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169 - 171
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72945

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-1168_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart