KEY001137358 Key Organics / BIONET

(Z)-N-[1-(4-chlorophenyl)ethylidene]hydroxylamine

  • MDL number MFCD00033237
  • Cas Number : 1956-39-4
  • Molecular Weight : 169.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 96 - 98
Iupac Name (1Z)-1-(4-chlorophenyl)ethanone oxime
Inchi Code 1S/C8H8ClNO/c1-6(10-11)7-2-4-8(9)5-3-7/h2-6H,1H3
Inchi Key FMWKNYZCWOFRTH-UHFFFAOYSA-N

Description

Melting Point 96 - 98
Purity 95%
Storage temp. Store under N2 at 4
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74167

Safety Information

signalword Warning
pcodes P411,P422, P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4F-002_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart