KEY001137495 Key Organics / BIONET

N-(2-hydroxyethyl)-2-[(3-methylphenyl)formamido]acetamide

  • MDL number MFCD00140963
  • Cas Number : 338419-52-6
  • Molecular Weight : 236.271

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109 - 112
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74333

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4F-910_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart