KEY001137586 Key Organics / BIONET

8-methoxy-2H-chromene-3-carbonitrile

  • MDL number MFCD00202017
  • Cas Number : 57543-69-8
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 105 - 106
Iupac Name 8-methoxy-2H-chromene-3-carbonitrile
Inchi Code 1S/C11H9NO2/c1-13-10-4-2-3-9-5-8(6-12)7-14-11(9)10/h2-5H,7H2,1H3
Inchi Key DAZAHWMZYLIXAU-UHFFFAOYSA-N

Description

Melting Point 105 - 106
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74440

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4G-318S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart