KEY001137595 Key Organics / BIONET

[1-methyl-3-(trifluoromethyl)-5-{[3-(trifluoromethyl)phenyl]sulfanyl}-1H-pyrazol-4-yl]methanol

  • MDL number MFCD00172636
  • Cas Number : 318239-53-1
  • Molecular Weight : 356.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61 - 62
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74453

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4G-335S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart