KEY001137633 Key Organics / BIONET

{5-[(4-fluorophenyl)sulfanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl}methanol

  • MDL number MFCD00172648
  • Cas Number : 318239-65-5
  • Molecular Weight : 306.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66 - 67
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74498

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4G-392S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart