KEY001137686 Key Organics / BIONET

ethyl 4-(4-fluorobenzoyl)-1,2-oxazole-3-carboxylate

  • MDL number MFCD00664758
  • Cas Number : 338420-60-3
  • Molecular Weight : 263.224

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73 - 75
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74562

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4G-950_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart