KEY001137695 Key Organics / BIONET

methyl (2E)-4-[(1-formylnaphthalen-2-yl)oxy]but-2-enoate

  • MDL number MFCD00231679
  • Cas Number : 174633-84-2
  • Molecular Weight : 270.284

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98 - 101
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74571

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4G-999_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart