KEY001137735 Key Organics / BIONET

2-[({[3-(trifluoromethyl)phenyl]carbamoyl}methyl)sulfanyl]acetic acid

  • MDL number MFCD00202305
  • Cas Number : 338421-05-9
  • Molecular Weight : 293.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 102 - 104
Iupac Name ({2-oxo-2-[3-(trifluoromethyl)anilino]ethyl}sulfanyl)acetic acid
Inchi Code 1S/C11H10F3NO3S/c12-11(13,14)7-2-1-3-8(4-7)15-9(16)5-19-6-10(17)18/h1-4H,5-6H2,(H,15,16)(H,17,18)
Inchi Key IJHVISSLZSLQOZ-UHFFFAOYSA-N

Description

Melting Point 102 - 104
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74616

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4H-311S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart