KEY001137755 Key Organics / BIONET

2-[({[(4-chlorophenyl)methyl]carbamoyl}methyl)sulfanyl]acetic acid

  • MDL number MFCD00202308
  • Cas Number : 338421-25-3
  • Molecular Weight : 273.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144 - 147
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74646

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4H-357S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart