KEY001137774 Key Organics / BIONET

2-[({[(4-methoxyphenyl)methyl]carbamoyl}methyl)sulfanyl]acetic acid

  • MDL number MFCD00202395
  • Cas Number : 338421-45-7
  • Molecular Weight : 269.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 92 - 94
Iupac Name ({2-[(4-methoxybenzyl)amino]-2-oxoethyl}sulfanyl)acetic acid
Inchi Code 1S/C12H15NO4S/c1-17-10-4-2-9(3-5-10)6-13-11(14)7-18-8-12(15)16/h2-5H,6-8H2,1H3,(H,13,14)(H,15,16)
Inchi Key YPXMMDXZXALGKO-UHFFFAOYSA-N

Description

Melting Point 92 - 94
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74670

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4H-391S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart