KEY001137775 Key Organics / BIONET

2-[({[(4-methylphenyl)methyl]carbamoyl}methyl)sulfanyl]acetic acid

  • MDL number MFCD00202396
  • Cas Number : 338421-46-8
  • Molecular Weight : 253.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131 - 133
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74672

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4H-393S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart