KEY001137874 Key Organics / BIONET

5-(methylamino)-3-(morpholin-4-yl)-1,2-thiazole-4-carbonitrile

  • MDL number MFCD00243960
  • Cas Number : 338422-41-6
  • Molecular Weight : 224.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176 - 178
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74778

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4H-913_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart