KEY001137877 Key Organics / BIONET

(2-{[(2,6-dichlorophenyl)methyl]sulfanyl}-1-methyl-1H-imidazol-5-yl)methanol

  • MDL number MFCD00215150
  • Cas Number : 338422-43-8
  • Molecular Weight : 303.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139 - 141
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74781

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4H-916_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart