KEY001137881 Key Organics / BIONET

(2-{[(5-chloro-1,2,3-thiadiazol-4-yl)methyl]sulfanyl}-1-methyl-1H-imidazol-5-yl)methanol

  • MDL number MFCD00232307
  • Cas Number : 338422-47-2
  • Molecular Weight : 276.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131 - 133
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74786

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4H-922_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart