KEY001137883 Key Organics / BIONET

1-(4-chlorophenyl)-2-{[5-(hydroxymethyl)-1-methyl-1H-imidazol-2-yl]sulfanyl}ethan-1-one

  • MDL number MFCD00215154
  • Cas Number : 338422-49-4
  • Molecular Weight : 296.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 115 - 118
Iupac Name 1-(4-chlorophenyl)-2-{[5-(hydroxymethyl)-1-methyl-1H-imidazol-2-yl]sulfanyl}ethanone
Inchi Code 1S/C13H13ClN2O2S/c1-16-11(7-17)6-15-13(16)19-8-12(18)9-2-4-10(14)5-3-9/h2-6,17H,7-8H2,1H3
Inchi Key LEOJWIQSSCPWPS-UHFFFAOYSA-N

Description

Melting Point 115 - 118
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74788

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4H-924_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart