KEY001137890 Key Organics / BIONET

methyl 2-(4-hydroxy-1,2-oxazolidine-2-carbonyl)benzoate

  • MDL number MFCD00202136
  • Cas Number : 80041-99-2
  • Molecular Weight : 251.238

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105 - 107
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74795

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4H-936_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart