KEY001137898 Key Organics / BIONET

(2-{[2-(4-methoxyphenoxy)ethyl]sulfanyl}-1-methyl-1H-imidazol-5-yl)methanol

  • MDL number MFCD00215158
  • Cas Number : 338422-59-6
  • Molecular Weight : 294.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124 - 126
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74804

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4H-947_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart