KEY001137911 Key Organics / BIONET

methyl 1-[(3,4-dichlorophenyl)carbamoyl]-4-hydroxypyrrolidine-2-carboxylate

  • MDL number MFCD00202783
  • Cas Number : 318247-31-3
  • Molecular Weight : 333.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175 - 177
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74818

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4H-966_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart