KEY001137980 Key Organics / BIONET

5-(4-chlorophenyl)-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD00231524
  • Cas Number : 35982-98-0
  • Molecular Weight : 230.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 281 - 284
Purity 95
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74898

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/4J-342S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart