KEY001138150 Key Organics / BIONET

N-(1-cyano-4-oxo-4H-quinolizin-3-yl)benzamide

  • MDL number MFCD00793861
  • Cas Number : 154411-16-2
  • Molecular Weight : 289.294

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 226 - 228
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75089

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4J-905_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart