KEY001138226 Key Organics / BIONET

5-[(4-bromophenyl)sulfanyl]-1-methyl-3-phenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD00244934
  • Cas Number : 318247-52-8
  • Molecular Weight : 373.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150 - 153
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75173

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4K-320S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart