KEY001138229 Key Organics / BIONET

[1-methyl-3-phenyl-5-(phenylsulfanyl)-1H-pyrazol-4-yl]methanol

  • MDL number MFCD00244937
  • Cas Number : 318247-55-1
  • Molecular Weight : 296.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134 - 136
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75176

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4K-323S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart