KEY001138359 Key Organics / BIONET

2-[2-(4-chlorophenyl)-4-phenyl-1,3-thiazol-5-yl]acetic acid

  • MDL number MFCD00663991
  • Cas Number : 23821-72-9
  • Molecular Weight : 329.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163 - 165
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75319

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4K-623S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart