KEY001138425 Key Organics / BIONET

potassium {4-[(E)-2-(thiophen-2-yl)ethenyl]pyrimidin-2-yl}sulfanide

  • MDL number MFCD00663706
  • Cas Number : 318248-61-2
  • Molecular Weight : 258.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name potassium 4-[(E)-2-(2-thienyl)ethenyl]-2-pyrimidinethiolate
Inchi Code 1S/C10H8N2S2.K/c13-10-11-6-5-8(12-10)3-4-9-2-1-7-14-9;/h1-7H,(H,11,12,13);/q;+1/p-1/b4-3+;
Inchi Key PWOMJJHEUSBNKS-BJILWQEISA-M

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75390

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4L-037_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart