KEY001138783 Key Organics / BIONET

1-[(2,4-dichlorophenyl)methyl]-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carbonitrile

  • MDL number MFCD00975040
  • Cas Number : 338748-72-4
  • Molecular Weight : 295.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144 - 146
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75771

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4M-920_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart