KEY001138793 Key Organics / BIONET

methyl 1-[(4-chlorophenyl)methyl]-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate

  • MDL number MFCD00975047
  • Cas Number : 338748-81-5
  • Molecular Weight : 293.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84 - 86
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75781

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4M-934_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart