KEY001138807 Key Organics / BIONET

4-chloro-N-(5-iodopyridin-2-yl)butanamide

  • MDL number MFCD01315195
  • Cas Number : 338748-92-8
  • Molecular Weight : 324.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 132 - 134
Iupac Name 4-chloro-N-(5-iodo-2-pyridinyl)butanamide
Inchi Code 1S/C9H10ClIN2O/c10-5-1-2-9(14)13-8-4-3-7(11)6-12-8/h3-4,6H,1-2,5H2,(H,12,13,14)
Inchi Key VDPISDJJUHBZDW-UHFFFAOYSA-N

Description

Melting Point 132 - 134
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75795

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/4M-954_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart