KEY001139118 Key Organics / BIONET

methyl 2-acetamidothiophene-3-carboxylate

  • MDL number MFCD00728968
  • Cas Number : 22288-81-9
  • Molecular Weight : 199.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103 - 105
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76121

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4N-802_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart