KEY001139183 Key Organics / BIONET

2-(2-formylphenoxy)benzonitrile

  • MDL number MFCD02082410
  • Cas Number : 451485-74-8
  • Molecular Weight : 223.231

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72 - 74
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76200

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4P-096_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart