KEY001139445 Key Organics / BIONET

2-(benzenesulfonyl)pyridin-3-amine

  • MDL number MFCD02186480
  • Cas Number : 444087-50-7
  • Molecular Weight : 234.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172 - 174
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76527

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-0608_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart