KEY001139710 Key Organics / BIONET

2-[(5-amino-1H-1,2,4-triazol-3-yl)sulfanyl]acetonitrile

  • MDL number MFCD00138376
  • Cas Number : 338751-47-6
  • Molecular Weight : 155.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130 - 133
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77673

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5D-007_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart