KEY001139786 Key Organics / BIONET

2-(6-chloropyridazin-3-yl)-2-phenylacetonitrile

  • MDL number MFCD00140699
  • Cas Number : 73535-73-6
  • Molecular Weight : 229.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128 - 130
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77750

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5D-128_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart