KEY001139887 Key Organics / BIONET

1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-ol

  • MDL number MFCD00139833
  • Cas Number : 122431-37-2
  • Molecular Weight : 166.103

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178 - 180
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77859

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5E-328S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart