KEY001139963 Key Organics / BIONET

3,3-bis(hydroxymethyl)-3,4-dihydro-2H-1-benzothiopyran-4-one

  • MDL number MFCD00139867
  • Cas Number : 29107-30-0
  • Molecular Weight : 224.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142 - 145
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77960

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5E-912_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart