KEY001140121 Key Organics / BIONET

ethyl 2-(5-phenyl-2H-1,2,3,4-tetrazol-2-yl)acetate

  • MDL number MFCD00268800
  • Cas Number : 21054-65-9
  • Molecular Weight : 232.243

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83 - 85
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78175

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5F-970_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart