KEY001140171 Key Organics / BIONET

6-methoxy-2H-chromene-3-carboxylic acid

  • MDL number MFCD00202057
  • Cas Number : 57543-62-1
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 190 - 192
Iupac Name 6-methoxy-2H-chromene-3-carboxylic acid
Inchi Code 1S/C11H10O4/c1-14-9-2-3-10-7(5-9)4-8(6-15-10)11(12)13/h2-5H,6H2,1H3,(H,12,13)
Inchi Key VOOCQPOSPBMQSK-UHFFFAOYSA-N

Description

Melting Point 190 - 192
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78230

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5G-302S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart