KEY001140175 Key Organics / BIONET

[1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol

  • MDL number MFCD00172652
  • Cas Number : 318469-22-6
  • Molecular Weight : 272.227

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80 - 82
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78235

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5G-308S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart