KEY001140177 Key Organics / BIONET

[5-(4-fluorobenzenesulfonyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol

  • MDL number MFCD00172654
  • Cas Number : 318469-24-8
  • Molecular Weight : 338.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127 - 129
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78238

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5G-311S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart