KEY001140203 Key Organics / BIONET

[1-methyl-3-(trifluoromethyl)-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazol-4-yl]methanol

  • MDL number MFCD00172663
  • Cas Number : 318469-40-8
  • Molecular Weight : 340.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 81 - 83
Iupac Name {1-methyl-3-(trifluoromethyl)-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazol-4-yl}methanol
Inchi Code 1S/C13H10F6N2O2/c1-21-11(9(6-22)10(20-21)13(17,18)19)23-8-4-2-3-7(5-8)12(14,15)16/h2-5,22H,6H2,1H3
Inchi Key BJRDDNWYIXOJIN-UHFFFAOYSA-N

Description

Melting Point 81 - 83
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78273

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5G-355S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart