KEY001140233 Key Organics / BIONET

ethyl 5-[(4-chlorophenyl)sulfanyl]-1,2,3-thiadiazole-4-carboxylate

  • MDL number MFCD00794804
  • Cas Number : 338760-67-1
  • Molecular Weight : 300.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91 - 92
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78308

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5G-398S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart