KEY001140272 Key Organics / BIONET

ethyl 4-[(4-chlorophenyl)carbamoyl]-5-methyl-1,2-oxazole-3-carboxylate

  • MDL number MFCD00664765
  • Cas Number : 134889-04-6
  • Molecular Weight : 308.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 108 - 110
Iupac Name ethyl 4-[(4-chloroanilino)carbonyl]-5-methyl-3-isoxazolecarboxylate
Inchi Code 1S/C14H13ClN2O4/c1-3-20-14(19)12-11(8(2)21-17-12)13(18)16-10-6-4-9(15)5-7-10/h4-7H,3H2,1-2H3,(H,16,18)
Inchi Key WMOOOFRTGANMKG-UHFFFAOYSA-N

Description

Melting Point 108 - 110
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78368

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5G-913_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart