KEY001140296 Key Organics / BIONET

ethyl 4-(pyridine-2-carbonyl)-1,2-oxazole-3-carboxylate

  • MDL number MFCD00664774
  • Cas Number : 338761-70-9
  • Molecular Weight : 246.222

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61 - 63
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78394

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5G-945_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart