KEY001140449 Key Organics / BIONET

4-[(5-chloro-1,2,3-thiadiazol-4-yl)methoxy]benzaldehyde

  • MDL number MFCD00232349
  • Cas Number : 338954-22-6
  • Molecular Weight : 254.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 79 - 81
Iupac Name 4-[(5-chloro-1,2,3-thiadiazol-4-yl)methoxy]benzaldehyde
Inchi Code 1S/C10H7ClN2O2S/c11-10-9(12-13-16-10)6-15-8-3-1-7(5-14)2-4-8/h1-5H,6H2
Inchi Key BUCQKDJJDPGCLD-UHFFFAOYSA-N

Description

Melting Point 79 - 81
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78578

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5H-921_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart