KEY001140485 Key Organics / BIONET

ethyl 6-fluoro-4-hydroxyquinoline-3-carboxylate

  • MDL number MFCD00173345
  • Cas Number : 318-35-4
  • Molecular Weight : 235.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 320 - 323
Iupac Name ethyl 6-fluoro-4-oxo-1,4-dihydro-3-quinolinecarboxylate
Inchi Code 1S/C12H10FNO3/c1-2-17-12(16)9-6-14-10-4-3-7(13)5-8(10)11(9)15/h3-6H,2H2,1H3,(H,14,15)
Inchi Key LNPRCADPRULVTR-UHFFFAOYSA-N

Description

Melting Point 320 - 323
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78615

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5J-031_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart