KEY001140521 Key Organics / BIONET

2-chloro-5-(4-chlorophenyl)pyridine-3-carbonitrile

  • MDL number MFCD00231583
  • Cas Number : 35982-99-1
  • Molecular Weight : 249.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182 - 184
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78652

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5J-312S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart